siRNA Details
| Gene Symbol: Brd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100903.1 | NP_001094373.1 | 6 | 1240 | ||
| XM_006241072.2 | XP_006241134.1 | 6 | 1244 | |||
| XM_006241073.2 | XP_006241135.1 | 6 | 1448 | |||
| XM_006241074.2 | XP_006241136.1 | 6 | 2172 | |||
| XM_008765170.1 | XP_008763392.1 | 5 | 2963 | |||
| XM_008765171.1 | XP_008763393.1 | 5 | 2963 | |||
| XM_008765172.1 | XP_008763394.1 | 5 | 2963 | |||
| XM_008765173.1 | XP_008763395.1 | 4 | 1490 | |||
| GenBank mRNA | BC100641.1 | - | 6 | 1187 | ||

Brd4
Genomic Map