siRNA Details
| Gene Symbol: Arhgap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012198.1 | NP_001012198.1 | 12, 13 | 1046 | ||
| NM_001080789.1 | NP_001074258.1 | 13, 14 | 1678 | |||
| XM_006241464.2 | XP_006241526.1 | 13, 14 | 1765 | |||
| XM_006241465.2 | XP_006241527.1 | 13, 14 | 1756 | |||
| XM_006241466.2 | XP_006241528.1 | 13, 14 | 1799 | |||
| XM_006241467.2 | XP_006241529.1 | 13, 14 | 1809 | |||
| XM_006241468.2 | XP_006241530.1 | 12, 13 | 1103 | |||
| GenBank mRNA | BC079334.1 | - | 12, 13 | 1046 | 7097179 | |
| BC107938.1 | - | 13, 14 | 1678 | 7378387 | ||

Arhgap9
Genomic Map