siRNA Details
| Gene Symbol: Arhgap9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012198.1 | NP_001012198.1 | 13 | 1155 | ||
| NM_001080789.1 | NP_001074258.1 | 14 | 1787 | |||
| XM_006241464.2 | XP_006241526.1 | 14 | 1874 | |||
| XM_006241465.2 | XP_006241527.1 | 14 | 1865 | |||
| XM_006241466.2 | XP_006241528.1 | 14 | 1908 | |||
| XM_006241467.2 | XP_006241529.1 | 14 | 1918 | |||
| XM_006241468.2 | XP_006241530.1 | 13 | 1212 | |||
| GenBank mRNA | BC079334.1 | - | 13 | 1155 | 7097179 | |
| BC107938.1 | - | 14 | 1787 | 7378387 | ||

Arhgap9
Genomic Map