siRNA Details
| Gene Symbol: Adck5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135798.1 | NP_001129270.1 | 15 | 1777 | ||
| XM_006241827.2 | XP_006241889.1 | 15 | 1681 | |||
| GenBank mRNA | BC168693.1 | - | 15 | 1754 | 7935927 | |
| BP468309.1 | - | 1 | 210 | |||

Adck5
Genomic Map