siRNA Details
| Gene Symbol: LOC102555083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006241920.2 | XP_006241982.1 | 1919 | |||
| Gene Symbol: Zfp647 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_343279.7 | XP_343280.3 | 5 | 1919 | ||

LOC102555083
Genomic Map