siRNA Details
Gene Symbol: Zcrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034940.1 | NP_001030112.1 | 2 | 152 | ||
GenBank mRNA | BC099747.1 | - | 2 | 152 | 5598773 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zcrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034940.1 | NP_001030112.1 | 2 | 152 | ||
GenBank mRNA | BC099747.1 | - | 2 | 152 | 5598773 |