siRNA Details
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122781.3 | NP_001116253.3 | 8 | 986 | ||
XM_006242354.2 | XP_006242416.1 | 8 | 1049 | |||
GenBank mRNA | BC159426.1 | - | 8 | 967 | 9034024 | |
DN933141.1 | - | 2 | 208 |
