siRNA Details
| Gene Symbol: Uhrf1bp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108753.1 | NP_001102223.1 | 10 | 1463 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uhrf1bp1l| Gene Symbol: Uhrf1bp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108753.1 | NP_001102223.1 | 10 | 1463 | ||