siRNA Details
| Gene Symbol: Taf1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014207.1 | NP_001014229.1 | 1 | 44 | ||
| XM_008765941.1 | XP_008764163.1 | 1 | 961 | |||
| XM_008765942.1 | XP_008764164.1 | 2 | 124 | |||
| XM_008765943.1 | XP_008764165.1 | 2 | 416 | |||
| XM_008765944.1 | XP_008764166.1 | 1 | 961 | |||
| XM_008765945.1 | XP_008764167.1 | 1 | 961 | |||
| XM_008765946.1 | XP_008764168.1 | 1 | 961 | |||
| XM_008765947.1 | XP_008764169.1 | 1 | 961 | |||
| XM_008765948.1 | XP_008764170.1 | 1 | 961 | |||
| GenBank mRNA | BC087647.1 | - | 1 | 44 | 7311303 | |

Taf1d
Genomic Map