siRNA Details
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108754.1 | NP_001102224.1 | 8 | 903 | ||
GenBank mRNA | FQ226445.1 | - | 8 | 769 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108754.1 | NP_001102224.1 | 8 | 903 | ||
GenBank mRNA | FQ226445.1 | - | 8 | 769 |