siRNA Details
Gene Symbol: Rp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108756.1 | NP_001102226.1 | 3 | 345 | ||
XM_008765960.1 | XP_008764182.1 | 3 | 439 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108756.1 | NP_001102226.1 | 3 | 345 | ||
XM_008765960.1 | XP_008764182.1 | 3 | 439 |