siRNA Details
| Gene Symbol: Dis3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008380.1 | NP_001008381.1 | 6, 7 | 829 | ||
| XM_006243261.2 | XP_006243323.1 | 6, 7 | 932 | |||
| XM_006243262.2 | XP_006243324.1 | 6, 7 | 736 | |||
| XM_006243263.2 | XP_006243325.1 | 6, 7 | 1261 | |||
| XM_008766311.1 | XP_008764533.1 | 6, 7 | 932 | |||
| GenBank mRNA | BC085932.1 | - | 6, 7 | 829 | 7112012 | |

Dis3l
Genomic Map