siRNA Details
| Gene Symbol: Myzap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014211.1 | NP_001014233.1 | 10, 11 | 1121 | ||
| XM_008766312.1 | XP_008764534.1 | 11, 12 | 1307 | |||
| GenBank mRNA | BC089899.1 | - | 10, 11 | 1121 | 7382871 | |

Myzap
Genomic Map