siRNA Details
| Gene Symbol: Dyx1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007010.1 | NP_001007011.1 | 6, 7 | 982 | ||
| XM_006243328.2 | XP_006243390.1 | 6, 7 | 1062 | |||
| XM_006243329.2 | XP_006243391.1 | 6, 7 | 1062 | |||
| XM_006243331.2 | XP_006243393.1 | 4, 5 | 702 | |||
| XM_008766313.1 | XP_008764535.1 | 6, 7 | 960 | |||
| XM_008766314.1 | XP_008764536.1 | 6, 7 | 791 | |||
| GenBank mRNA | AY428506.1 | - | 6, 7 | 982 | ||
| BC085838.1 | - | 6, 7 | 926 | 7135744 | ||

Dyx1c1
Genomic Map