siRNA Details
| Gene Symbol: Slc9a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271438.1 | NP_001258367.1 | 579 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a9| Gene Symbol: Slc9a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271438.1 | NP_001258367.1 | 579 | |||