siRNA Details
| Gene Symbol: Xpo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108789.3 | NP_001102259.2 | 23 | 2638 | ||
| GenBank mRNA | DN935088.1 | - | 6 | 602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpo5| Gene Symbol: Xpo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108789.3 | NP_001102259.2 | 23 | 2638 | ||
| GenBank mRNA | DN935088.1 | - | 6 | 602 | ||