siRNA Details
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013225.2 | NP_001013243.2 | 12 | 1777 | ||
XM_006244543.2 | XP_006244605.1 | 12 | 1819 | |||
XM_006244544.2 | XP_006244606.1 | 12 | 1609 | |||
XM_006244545.2 | XP_006244607.1 | 11 | 3693 | |||
XM_006244546.2 | XP_006244608.1 | 12 | 1598 | |||
GenBank mRNA | BC088339.1 | - | 1 | 372 |
