siRNA Details
| Gene Symbol: Tmem63b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750650.3 | XP_003750698.1 | 16 | 1635 | ||
| XM_006244582.2 | XP_006244644.1 | 15 | 1597 | |||
| XM_006244583.1 | XP_006244645.1 | 15 | 1677 | |||
| XM_008766903.1 | XP_008765125.1 | 16 | 1625 | |||

Tmem63b
Genomic Map