siRNA Details
Gene Symbol: Tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108795.1 | NP_001102265.1 | 3 | 802 | ||
XM_008767117.1 | XP_008765339.1 | 3 | 797 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmeff2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108795.1 | NP_001102265.1 | 3 | 802 | ||
XM_008767117.1 | XP_008765339.1 | 3 | 797 |