siRNA Details
| Gene Symbol: Col4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135759.1 | NP_001129231.1 | 49 | 4496 | ||
| XM_006245259.2 | XP_006245321.1 | 49 | 4719 | |||
| GenBank mRNA | L47281.2 | - | 2 | 176 | ||
| Gene Symbol: LOC100911572 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750719.3 | XP_003750767.2 | 8 | 1311 | ||

Col4a3
Genomic Map