siRNA Details
| Gene Symbol: Rhox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135607.1 | NP_001129079.1 | 397 | |||
| XM_008773382.1 | XP_008771604.1 | 198 | ||||
| XM_008773383.1 | XP_008771605.1 | 2, 3 | 144 | |||
| GenBank mRNA | DQ058651.1 | - | 397 | |||
| Gene Symbol: LOC103690001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257477.2 | XP_006257539.1 | 2, 3 | 144 | ||
| XM_008773510.1 | XP_008771732.1 | 2, 3 | 198 | |||

Rhox3
Genomic Map