siRNA Details
| Gene Symbol: Tekt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014224.2 | NP_001014246.1 | 1 | 158 | ||
| GenBank mRNA | AB294402.1 | - | 1 | 158 | ||
| BC079040.1 | - | 1 | 230 | 7114771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tekt5| Gene Symbol: Tekt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014224.2 | NP_001014246.1 | 1 | 158 | ||
| GenBank mRNA | AB294402.1 | - | 1 | 158 | ||
| BC079040.1 | - | 1 | 230 | 7114771 | ||