siRNA Details
| Gene Symbol: RGD1562673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108823.1 | NP_001102293.1 | 2 | 140 | ||
| XM_008767589.1 | XP_008765811.1 | 3 | 1073 | |||
| GenBank mRNA | BC167085.1 | - | 480 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562673| Gene Symbol: RGD1562673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108823.1 | NP_001102293.1 | 2 | 140 | ||
| XM_008767589.1 | XP_008765811.1 | 3 | 1073 | |||
| GenBank mRNA | BC167085.1 | - | 480 | |||