siRNA Details
Gene Symbol: RGD1562673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108823.1 | NP_001102293.1 | 3 | 397 | ||
XM_008767589.1 | XP_008765811.1 | 4 | 1330 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1562673 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108823.1 | NP_001102293.1 | 3 | 397 | ||
XM_008767589.1 | XP_008765811.1 | 4 | 1330 |