siRNA Details
| Gene Symbol: Cnot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008382.1 | NP_001008383.1 | 5 | 666 | ||
| XM_006246392.2 | XP_006246454.1 | 4 | 725 | |||
| GenBank mRNA | BC085856.1 | - | 5 | 666 | 7190112 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnot8| Gene Symbol: Cnot8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008382.1 | NP_001008383.1 | 5 | 666 | ||
| XM_006246392.2 | XP_006246454.1 | 4 | 725 | |||
| GenBank mRNA | BC085856.1 | - | 5 | 666 | 7190112 | |