siRNA Details
| Gene Symbol: Natd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170541.1 | NP_001164012.1 | 2 | 139 | ||
| GenBank mRNA | CB546029.1 | - | 2 | 113 | ||
| CK480689.1 | - | 2 | 139 | |||
| FQ231549.1 | - | 2 | 181 | |||
| Gene Symbol: LOC100911674 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750836.3 | XP_003750884.1 | 2 | 869 | ||

Natd1
Genomic Map