siRNA Details
| Gene Symbol: Grap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025749.1 | NP_001020920.1 | 4 | 490 | ||
| GenBank mRNA | BC098740.1 | - | 4 | 490 | 7386199 | |
| FQ228943.1 | - | 4 | 457 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grap| Gene Symbol: Grap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025749.1 | NP_001020920.1 | 4 | 490 | ||
| GenBank mRNA | BC098740.1 | - | 4 | 490 | 7386199 | |
| FQ228943.1 | - | 4 | 457 | |||