siRNA Details
| Gene Symbol: Rnf222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108828.1 | NP_001102298.1 | 3 | 633 | ||
| XM_006246764.2 | XP_006246826.1 | 5 | 1150 | |||
| XM_006246765.2 | XP_006246827.1 | 4 | 3632 | |||
| XM_008767845.1 | XP_008766067.1 | 3 | 3506 | |||
| XM_008767846.1 | XP_008766068.1 | 5 | 2986 | |||
| XM_008767847.1 | XP_008766069.1 | 3 | 933 | |||

Rnf222
Genomic Map