siRNA Details
| Gene Symbol: Glod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014227.1 | NP_001014249.1 | 8 | 830 | ||
| GenBank mRNA | BC088458.1 | - | 8 | 830 | 7127323 | |
| FQ210234.1 | - | 8 | 848 | |||
| FQ230415.1 | - | 8 | 806 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glod4| Gene Symbol: Glod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014227.1 | NP_001014249.1 | 8 | 830 | ||
| GenBank mRNA | BC088458.1 | - | 8 | 830 | 7127323 | |
| FQ210234.1 | - | 8 | 848 | |||
| FQ230415.1 | - | 8 | 806 | |||