siRNA Details
| Gene Symbol: Eral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013229.1 | NP_001013247.1 | 5 | 587 | ||
| XM_008768087.1 | XP_008766309.1 | 5 | 583 | |||
| GenBank mRNA | BC089885.1 | - | 5 | 587 | 7370040 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eral1| Gene Symbol: Eral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013229.1 | NP_001013247.1 | 5 | 587 | ||
| XM_008768087.1 | XP_008766309.1 | 5 | 583 | |||
| GenBank mRNA | BC089885.1 | - | 5 | 587 | 7370040 | |