siRNA Details
| Gene Symbol: Eral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013229.1 | NP_001013247.1 | 1 | 106 | ||
| XM_008768087.1 | XP_008766309.1 | 1 | 102 | |||
| GenBank mRNA | BC089885.1 | - | 1 | 106 | 7370040 | |
| Gene Symbol: LOC103693394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768143.1 | XP_008766365.1 | 1 | 497 | ||

Eral1
Genomic Map