siRNA Details
| Gene Symbol: Cct6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014228.1 | NP_001014250.1 | 6 | 761 | ||
| GenBank mRNA | BC079020.1 | - | 6 | 761 | 7113829 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cct6b| Gene Symbol: Cct6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014228.1 | NP_001014250.1 | 6 | 761 | ||
| GenBank mRNA | BC079020.1 | - | 6 | 761 | 7113829 | |