siRNA Details
| Gene Symbol: Map2k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025425.1 | NP_001020596.1 | 7 | 802 | ||
| XM_006248768.2 | XP_006248830.1 | 8 | 987 | |||
| XM_006248769.1 | XP_006248831.1 | 8 | 992 | |||
| XM_008768985.1 | XP_008767207.1 | 7 | 943 | |||
| GenBank mRNA | AY879265.1 | - | 7 | 802 | ||
| GU264001.1 | - | 8 | 850 | |||

Map2k7
Genomic Map