siRNA Details
| Gene Symbol: Rac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134366.1 | NP_599193.1 | 6 | 498 | ||
| XM_008768986.1 | XP_008767208.1 | 7 | 589 | |||
| GenBank mRNA | AY491395.1 | - | 6 | 498 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rac1| Gene Symbol: Rac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134366.1 | NP_599193.1 | 6 | 498 | ||
| XM_008768986.1 | XP_008767208.1 | 7 | 589 | |||
| GenBank mRNA | AY491395.1 | - | 6 | 498 | ||