siRNA Details
| Gene Symbol: Cnpy4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108852.2 | NP_001102322.1 | 5, 6 | 604 | ||
| GenBank mRNA | BC162041.1 | - | 5, 6 | 604 | 7100495 | |
| FQ226241.1 | - | 5, 6 | 623 | |||
| FQ229096.1 | - | 5, 6 | 649 | |||

Cnpy4
Genomic Map