siRNA Details
| Gene Symbol: Phlda3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012206.2 | NP_001012206.1 | 1 | 349 | ||
| GenBank mRNA | BC087038.1 | - | 1 | 335 | 7096388 | |
| EV766228.1 | - | 1 | 362 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phlda3| Gene Symbol: Phlda3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012206.2 | NP_001012206.1 | 1 | 349 | ||
| GenBank mRNA | BC087038.1 | - | 1 | 335 | 7096388 | |
| EV766228.1 | - | 1 | 362 | |||