siRNA Details
| Gene Symbol: Pigc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012207.1 | NP_001012207.1 | 2 | 856 | ||
| XM_006250182.2 | XP_006250244.1 | 2 | 1022 | |||
| XM_006250183.1 | XP_006250245.1 | 2 | 922 | |||
| XM_008769665.1 | XP_008767887.1 | 2 | 960 | |||
| GenBank mRNA | BC087080.1 | - | 2 | 856 | 7189037 | |

Pigc
Genomic Map