siRNA Details
| Gene Symbol: Vamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108856.1 | NP_001102326.1 | 4 | 255 | ||
| XM_006250184.2 | XP_006250246.1 | 4 | 342 | |||
| XM_006250186.2 | XP_006250248.1 | 4 | 334 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vamp4| Gene Symbol: Vamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108856.1 | NP_001102326.1 | 4 | 255 | ||
| XM_006250184.2 | XP_006250246.1 | 4 | 342 | |||
| XM_006250186.2 | XP_006250248.1 | 4 | 334 | |||