siRNA Details
| Gene Symbol: Pycr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012208.1 | NP_001012208.1 | 5, 6 | 746 | ||
| GenBank mRNA | BC079222.1 | - | 5, 6 | 746 | 7132148 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pycr2| Gene Symbol: Pycr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012208.1 | NP_001012208.1 | 5, 6 | 746 | ||
| GenBank mRNA | BC079222.1 | - | 5, 6 | 746 | 7132148 | |