siRNA Details
| Gene Symbol: Fbxo48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108863.1 | NP_001102333.1 | 1 | 275 | ||
| XM_006251518.2 | XP_006251580.1 | 3 | 407 | |||
| XM_008770433.1 | XP_008768655.1 | 2 | 1583 | |||
| XM_008770434.1 | XP_008768656.1 | 3 | 945 | |||

Fbxo48
Genomic Map