siRNA Details
| Gene Symbol: Cnrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014232.1 | NP_001014254.1 | 3 | 575 | ||
| GenBank mRNA | AY883936.1 | - | 3 | 664 | ||
| BC088754.1 | - | 3 | 575 | 7319918 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnrip1| Gene Symbol: Cnrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014232.1 | NP_001014254.1 | 3 | 575 | ||
| GenBank mRNA | AY883936.1 | - | 3 | 664 | ||
| BC088754.1 | - | 3 | 575 | 7319918 | ||