siRNA Details
| Gene Symbol: Lrrc63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024803.1 | NP_001019974.1 | 3 | 790 | ||
| XM_006252325.2 | XP_006252387.1 | 3 | 850 | |||
| XM_006252326.2 | XP_006252388.1 | 3 | 884 | |||
| XM_006252327.2 | XP_006252389.1 | 3 | 833 | |||
| GenBank mRNA | BC083605.1 | - | 3 | 850 | ||
| BC097406.1 | - | 3 | 790 | 7453994 | ||

Lrrc63
Genomic Map