siRNA Details
Gene Symbol: Ssbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014236.1 | NP_001014258.1 | 3, 4 | 333 | ||
XM_006252912.2 | XP_006252974.1 | 3, 4 | 346 | |||
XM_008771139.1 | XP_008769361.1 | 3, 4 | 348 | |||
XM_008771140.1 | XP_008769362.1 | 3, 4 | 346 | |||
XM_008771141.1 | XP_008769363.1 | 3, 4 | 346 | |||
XM_008771142.1 | XP_008769364.1 | 3, 4 | 346 | |||
GenBank mRNA | BC079147.1 | - | 3, 4 | 333 | 7128392 |
