siRNA Details
| Gene Symbol: Ssbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014236.1 | NP_001014258.1 | 5, 6 | 501 | ||
| XM_008771140.1 | XP_008769362.1 | 5, 6 | 514 | |||
| XM_008771142.1 | XP_008769364.1 | 5, 6 | 514 | |||
| GenBank mRNA | BC079147.1 | - | 5, 6 | 501 | 7128392 | |

Ssbp4
Genomic Map