siRNA Details
| Gene Symbol: Poteg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014238.1 | NP_001014260.1 | 1 | 234 | ||
| XM_008771330.1 | XP_008769552.1 | 1 | 283 | |||
| GenBank mRNA | BC079270.1 | - | 1 | 234 | 7133714 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Poteg| Gene Symbol: Poteg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014238.1 | NP_001014260.1 | 1 | 234 | ||
| XM_008771330.1 | XP_008769552.1 | 1 | 283 | |||
| GenBank mRNA | BC079270.1 | - | 1 | 234 | 7133714 | |