siRNA Details
| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108876.1 | NP_001102346.1 | 3 | 491 | ||
| GenBank mRNA | FQ216033.1 | - | 3 | 261 | ||
| FQ220497.1 | - | 3 | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lsm1| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108876.1 | NP_001102346.1 | 3 | 491 | ||
| GenBank mRNA | FQ216033.1 | - | 3 | 261 | ||
| FQ220497.1 | - | 3 | 359 | |||