siRNA Details
| Gene Symbol: Sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012213.1 | NP_001012213.1 | 2 | 195 | ||
| XM_006253645.2 | XP_006253707.1 | 2 | 406 | |||
| XM_008771502.1 | XP_008769724.1 | 2 | 167 | |||
| GenBank mRNA | BC078935.1 | - | 2 | 195 | 7108166 | |
| FQ209748.1 | - | 199 | ||||

Sfxn1
Genomic Map