siRNA Details
| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108883.1 | NP_001102353.1 | 3 | 732 | ||
| XM_008771877.1 | XP_008770099.1 | 3 | 862 | |||
| XM_008771878.1 | XP_008770100.1 | 3 | 1936 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Suv39h2| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108883.1 | NP_001102353.1 | 3 | 732 | ||
| XM_008771877.1 | XP_008770099.1 | 3 | 862 | |||
| XM_008771878.1 | XP_008770100.1 | 3 | 1936 | |||