siRNA Details
| Gene Symbol: Polr2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108886.1 | NP_001102356.1 | 2 | 161 | ||
| GenBank mRNA | FQ228021.1 | - | 2 | 132 | ||
| FQ233353.1 | - | 2 | 128 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr2d| Gene Symbol: Polr2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108886.1 | NP_001102356.1 | 2 | 161 | ||
| GenBank mRNA | FQ228021.1 | - | 2 | 132 | ||
| FQ233353.1 | - | 2 | 128 | |||