siRNA Details
| Gene Symbol: Chmp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108906.1 | NP_001102376.1 | 3 | 389 | ||
| XM_006228117.1 | XP_006228179.1 | 3 | 779 | |||
| XM_006228118.1 | XP_006228180.1 | 3 | 779 | |||
| GenBank mRNA | BC167093.1 | - | 3 | 387 | 9088608 | |
| FQ222002.1 | - | 3 | 355 | |||
| FQ229914.1 | - | 3 | 297 | |||
| FQ230074.1 | - | 3 | 390 | |||
| FQ230732.1 | - | 3 | 405 | |||
| FQ231086.1 | - | 3 | 410 | |||
| FQ231147.1 | - | 3 | 384 | |||
| FQ233974.1 | - | 3 | 300 | |||

Chmp2a
Genomic Map